missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TRIM10 Monoclonal Antibody (K92040_4A6), Invitrogen™

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA556015

 View more versions of this product

Catalog No. MA556015

Only null left
Explore available promotions

Description

Description

Sequence of this protein is as follows: LEESPTCPLC KEPFRPGSFR PNWQLANVVE NIERLQLVST LGLGEEDVCQ EHGEKIYFFC EDDEMQLCVV CREAGEHATH TMRFLEDAAA PYREQIHKCL KCLRKEREEI QEIQSRENKR

The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This protein localizes to cytoplasmic bodies. Studies in mice suggest that this protein plays a role in terminal differentiation of erythroid cells. Alternate splicing of this gene generates two transcript variants encoding different isoforms.
Specifications

Specifications

TRIM10
Monoclonal
1 mg/mL
PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Q9UDY6
TRIM10
Recombinant human TRIM10 (immunogen range is 51-170).
200 μL
Primary
Human, Rat
Antibody
IgG1
Western Blot
K92040_4A6
Unconjugated
TRIM10
AI324236; B30-RING finger protein; BB139825; Hematopoietic RING finger 1; Herf1; RFB30; RING finger protein 9; Rnf9; TRIM10; tripartite motif containing 10; tripartite motif protein 10; tripartite motif-containing 10; tripartite motif-containing protein 10; Zn-finger protein
Mouse
Protein A
RUO
10107, 294210
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions