missing translation for 'onlineSavingsMsg'
Learn More

Trilinolein 95.0+%, TCI America™

Catalog No. p-7136750
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T13885G 5 g
T138825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T13885G Supplier TCI America Supplier No. T13885G
Only null left

Chemical Identifiers

CAS 537-40-6
Molecular Formula C57H98O6
Molecular Weight (g/mol) 879.405
MDL Number MFCD00042909
InChI Key HBOQXIRUPVQLKX-BBWANDEASA-N
Synonym Glycerol Tri-9,12-octadecadienoate, 1,2,3-Tri(cis,cis-9,12-octadecadienoyl) Glycerol
PubChem CID 5322095
ChEBI CHEBI:75844
IUPAC Name 2,3-bis[[(9Z,12Z)-octadeca-9,12-dienoyl]oxy]propyl (9Z,12Z)-octadeca-9,12-dienoate
SMILES CCCCCC=CCC=CCCCCCCCC(=O)OCC(COC(=O)CCCCCCCC=CCC=CCCCCC)OC(=O)CCCCCCCC=CCC=CCCCCC

Specifications

Quantity 5 g
Formula Weight 879.41
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Trilinolein
TSCA No
Recommended Storage Refrigerator