missing translation for 'onlineSavingsMsg'
Learn More

Triisopropylsilylacetylene, 97%

Catalog No. p-7035682
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5340506 5 g
AAH5340514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5340506 Supplier Thermo Scientific Chemicals Supplier No. H5340506

CAS: 89343-06-6 | C11H22Si | 182.382 g/mol

Triisopropylsilylacetylene is used as a reagent for the rhodium-catalyzed asymmetric alkylation of alpha, beta-unsaturated carbonyl compounds as well as in the synthesis of beta-alkynylated nitroalkanes. It is utilized in the preparation of bromoethynyl-triisopropyl-silane in the presence of the reagents such as N-bromo succinimide and silver nitrate. It is actively in involved in Sonogashira coupling reaction with 1-bromo-3-iodo-5-tertbutylbenzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triisopropylsilylacetylene is used as a reagent for the rhodium-catalyzed asymmetric alkylation of alpha, beta-unsaturated carbonyl compounds as well as in the synthesis of beta-alkynylated nitroalkanes. It is utilized in the preparation of bromoethynyl-triisopropyl-silane in the presence of the reagents such as N-bromo succinimide and silver nitrate. It is actively in involved in Sonogashira coupling reaction with 1-bromo-3-iodo-5-tertbutylbenzene.

Solubility
Miscible with organic solvents.

Notes
Incompatible with strong acids, strong bases and strong oxidizing agents.

Chemical Identifiers

CAS 89343-06-6
Molecular Formula C11H22Si
Molecular Weight (g/mol) 182.382
MDL Number MFCD00075452
InChI Key KZGWPHUWNWRTEP-UHFFFAOYSA-N
Synonym triisopropylsilyl acetylene, ethynyltriisopropylsilane, triisopropylsilylacetylene, ethynyltris propan-2-yl silane, ethynyl-tri propan-2-yl silane, silane, ethynyltris 1-methylethyl, triisopropylsilylacetylen, acmc-209qzk, ethynyltriisopropyl-silane
PubChem CID 2734682
IUPAC Name ethynyl-tri(propan-2-yl)silane
SMILES CC(C)[Si](C#C)(C(C)C)C(C)C

Specifications

Boiling Point 50°C to 52°C (0.6mmHg)
Quantity 5 g
Linear Formula [(CH3)2CH]3SiC≡CH
UN Number UN1993
Beilstein 3536241
Flash Point 56°C (133°F)
Solubility Information Miscible with organic solvents.
Formula Weight 182.38
Percent Purity 97%
Refractive Index 1.4527
Density 0.813
Chemical Name or Material Triisopropylsilylacetylene
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only