missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triisopropylsilyl Trifluoromethanesulfonate 98.0+%, TCI America™
Supplier: TCI America T158825G
Specifications
| Triisopropylsilyl Trifluoromethanesulfonate | |
| 87°C | |
| MFCD00009913 | |
| 1760 | |
| LHJCZOXMCGQVDQ-UHFFFAOYSA-N | |
| tri(propan-2-yl)silyl trifluoromethanesulfonate | |
| 2724529 | |
| ≥98.0% (T) |
| 80522-42-5 | |
| C10H21F3O3SSi | |
| 25 g | |
| triisopropylsilyl trifluoromethanesulfonate, triisopropylsilyl triflate, triisopropylsilyltrifluoromethanesulfonate, trifluoromethanesulfonic acid triisopropylsilyl ester, tips-otf, tips triflate, wacker silane ip3-triflate, triisopropylsilyl trifluoromethanesulphonate, triisopropylsilyl-trifluoromethanesulfonate, methanesulfonic acid, trifluoro-, tris 1-methylethyl silyl ester | |
| CC(C)[Si](C(C)C)(C(C)C)OS(=O)(=O)C(F)(F)F | |
| 306.415 | |
| 306.42 | |
| Liquid |
Chemical Identifiers
| 80522-42-5 | |
| 306.415 | |
| LHJCZOXMCGQVDQ-UHFFFAOYSA-N | |
| 2724529 | |
| CC(C)[Si](C(C)C)(C(C)C)OS(=O)(=O)C(F)(F)F |
| C10H21F3O3SSi | |
| MFCD00009913 | |
| triisopropylsilyl trifluoromethanesulfonate, triisopropylsilyl triflate, triisopropylsilyltrifluoromethanesulfonate, trifluoromethanesulfonic acid triisopropylsilyl ester, tips-otf, tips triflate, wacker silane ip3-triflate, triisopropylsilyl trifluoromethanesulphonate, triisopropylsilyl-trifluoromethanesulfonate, methanesulfonic acid, trifluoro-, tris 1-methylethyl silyl ester | |
| tri(propan-2-yl)silyl trifluoromethanesulfonate |
Safety and Handling
TSCA : No