accessibility menu, dialog, popup

UserName

CH3H3CSiCH3H3CCH3H3COSOOFFF

CAS RN 80522-42-5

IUPAC Name: tris(propan-2-yl)silyl trifluoromethanesulfonate
Synonyms: triisopropylsilyl trifluoromethanesulfonate
Molecular Weight (g/mol): 306.42
Molecular Formula: C10H21F3O3SSi
InChi Key: LHJCZOXMCGQVDQ-UHFFFAOYSA-N
SMILES: CC(C)[Si](OS(=O)(=O)C(F)(F)F)(C(C)C)C(C)C
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (5)
  • (2)
  • (2)
  • (5)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (4)