missing translation for 'onlineSavingsMsg'
Learn More

Triisopropylsilyl Trifluoromethanesulfonate 98.0+%, TCI America™

Catalog No. T15885G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15885G 5 g
T158825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T15885G Supplier TCI America Supplier No. T15885G

Chemical Identifiers

CAS 80522-42-5
Molecular Formula C10H21F3O3SSi
Molecular Weight (g/mol) 306.415
MDL Number MFCD00009913
InChI Key LHJCZOXMCGQVDQ-UHFFFAOYSA-N
Synonym triisopropylsilyl trifluoromethanesulfonate, triisopropylsilyl triflate, triisopropylsilyltrifluoromethanesulfonate, trifluoromethanesulfonic acid triisopropylsilyl ester, tips-otf, tips triflate, wacker silane ip3-triflate, triisopropylsilyl trifluoromethanesulphonate, triisopropylsilyl-trifluoromethanesulfonate, methanesulfonic acid, trifluoro-, tris 1-methylethyl silyl ester
PubChem CID 2724529
IUPAC Name tri(propan-2-yl)silyl trifluoromethanesulfonate
SMILES CC(C)[Si](C(C)C)(C(C)C)OS(=O)(=O)C(F)(F)F

Specifications

Boiling Point 87°C
Quantity 5 g
UN Number 1760
Formula Weight 306.42
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Triisopropylsilyl Trifluoromethanesulfonate