missing translation for 'onlineSavingsMsg'
Learn More

Triisopropylsilyl-trifluoromethanesulfonate, 97%

Catalog No. AC250700100
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC250700100 10 g Glass bottle
AC250700500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC250700100 Supplier Thermo Scientific Chemicals Supplier No. 250700100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 80522-42-5 | C10H21F3O3SSi | 306.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 80522-42-5
Molecular Formula C10H21F3O3SSi
Molecular Weight (g/mol) 306.42
MDL Number MFCD00009913
InChI Key LHJCZOXMCGQVDQ-UHFFFAOYSA-N
Synonym triisopropylsilyl trifluoromethanesulfonate, triisopropylsilyl triflate, triisopropylsilyltrifluoromethanesulfonate, trifluoromethanesulfonic acid triisopropylsilyl ester, tips-otf, tips triflate, wacker silane ip3-triflate, triisopropylsilyl trifluoromethanesulphonate, triisopropylsilyl-trifluoromethanesulfonate, methanesulfonic acid, trifluoro-, tris 1-methylethyl silyl ester
PubChem CID 2724529
IUPAC Name tri(propan-2-yl)silyl trifluoromethanesulfonate
SMILES CC(C)[Si](C(C)C)(C(C)C)OS(=O)(=O)C(F)(F)F

Specifications

Color Brown to Brown-Orange
Density 1.1400g/mL
Flash Point 100°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.4130 to 1.415
Linear Formula CF3SO3Si(CH(CH3)2)3
Quantity 10 g
Fieser 13,11
Specific Gravity 1.14
Solubility Information Solubility in water: hydrolyzes
Formula Weight 306.42
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Triisopropylsilyl-trifluoromethanesulfonate, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

RUO – Research Use Only