missing translation for 'onlineSavingsMsg'
Learn More

Triisopropyl[(trimethylsilyl)ethynyl]silane 97.0+%, TCI America™

Catalog No. T32711ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T32711ML 1 mL
T32715ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T32711ML Supplier TCI America Supplier No. T32711ML
Only null left

Chemical Identifiers

CAS 107474-02-2
Molecular Formula C14H30Si2
Molecular Weight (g/mol) 254.564
InChI Key ZADMVBSHDANSRO-UHFFFAOYSA-N
PubChem CID 13779468
IUPAC Name trimethyl-[2-tri(propan-2-yl)silylethynyl]silane
SMILES CC(C)[Si](C#C[Si](C)(C)C)(C(C)C)C(C)C

Specifications

Color Yellow
Boiling Point 57°C
Quantity 1 mL
Formula Weight 254.56
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Triisopropyl[(trimethylsilyl)ethynyl]silane