accessibility menu, dialog, popup

UserName

CAS RN 107474-02-2

CH3CH3SiSiH3CCH3CH3H3CCH3H3CCH3

CAS RN 107474-02-2

IUPAC Name: trimethyl({2-[tris(propan-2-yl)silyl]ethynyl})silane
Synonyms: triisopropyl[2-(trimethylsilyl)ethynyl]silane
Molecular Weight (g/mol): 254.56
Molecular Formula: C14H30Si2
InChi Key: ZADMVBSHDANSRO-UHFFFAOYSA-N
SMILES: CC(C)[Si](C#C[Si](C)(C)C)(C(C)C)C(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results