missing translation for 'onlineSavingsMsg'
Learn More

Triisopropyl Borate 98.0+%, TCI America™

Catalog No. B0521500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0521500ML 500 mL
B052125ML 25 mL
B0521100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B0521500ML Supplier TCI America Supplier No. B0521500ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5419-55-6
Molecular Formula C9H21BO3
Molecular Weight (g/mol) 188.074
MDL Number MFCD00008872
InChI Key NHDIQVFFNDKAQU-UHFFFAOYSA-N
Synonym triisopropyl borate, isopropyl borate, triisopropoxyborane, triisopropoxyboron, boron isopropoxide, triisopropyl orthoborate, boric acid triisopropyl ester, triisopropoxy borane, boron triisopropoxide, boric acid h3bo3 , tris 1-methylethyl ester
PubChem CID 21531
IUPAC Name tripropan-2-yl borate
SMILES B(OC(C)C)(OC(C)C)OC(C)C

Specifications

Color Colorless
Boiling Point 140°C
Quantity 500 mL
UN Number 2616
Formula Weight 188.07
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Triisopropyl Borate
EINECSNumber (2)-2101
RTECSNumber ED5950000
TSCA Yes