missing translation for 'onlineSavingsMsg'
Learn More

Triisopropyl borate, 98+%

Catalog No. p-3737655
Change view
Click to view available options
Quantity:
80 g
750 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC188740800 80 g Glass bottle
AC188747500 750 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC188740800 Supplier Thermo Scientific Chemicals Supplier No. 188740800
Only null left

CAS: 5419-55-6 | C9H21BO3 | 188.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5419-55-6
Molecular Formula C9H21BO3
Molecular Weight (g/mol) 188.07
MDL Number MFCD00008872
InChI Key NHDIQVFFNDKAQU-UHFFFAOYSA-N
Synonym triisopropyl borate, isopropyl borate, triisopropoxyborane, triisopropoxyboron, boron isopropoxide, triisopropyl orthoborate, boric acid triisopropyl ester, triisopropoxy borane, boron triisopropoxide, boric acid h3bo3 , tris 1-methylethyl ester
PubChem CID 21531
IUPAC Name tripropan-2-yl borate
SMILES B(OC(C)C)(OC(C)C)OC(C)C

Specifications

Melting Point -59°C
Color Colorless
Density 0.8140g/mL
Boiling Point 140°C
Flash Point 17°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.3755 to 1.3775
Linear Formula [(CH3)2CHO]3B
Quantity 80 g
Beilstein 01, 363
Specific Gravity 0.814
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in alcohol,ether and benzene
Formula Weight 188.07
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Triisopropyl borate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 226-529-9

RUO – Research Use Only