missing translation for 'onlineSavingsMsg'
Learn More

Triisopropanolamine cyclic borate, 98+%

CAS: 101-00-8 | C9H18BNO3 | 199.06 g/mol

$479.82 - $479.82

Chemical Identifiers

CAS 101-00-8
Molecular Formula C9H18BNO3
Molecular Weight (g/mol) 199.06
MDL Number MFCD03791298
InChI Key IWKGJTDSJPLUCE-UHFFFAOYSA-N
Synonym triisopropanolamine cyclic borate, 3,7,10-trimethyl-2,8,9-trioxa-5-aza-1-borabicyclo 3.3.3 undecane, borester 21, triisopropanolamine borate, boric acid, tris 1-amino-2-propyl ester, 2-propanol, 1,1',1-nitrilotri-, cyclic borate, 3,7,10-trimethyl-4,6,11-trioxa-1-aza-5-borabicyclo 3.3.3 undecane, 2,8,9-trioxa-5-aza-1-borabicyclo 3.3.3 undecane, 3,7,10-trimethyl, triisopropanolamineborate
PubChem CID 225550
IUPAC Name 3,7,10-trimethyl-4,6,11-trioxa-1-aza-5-borabicyclo[3.3.3]undecane
SMILES B12OC(CN(CC(O1)C)CC(O2)C)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC269241000
SDS View Documents
Thermo Scientific Chemicals
269241000
100 g Glass bottle
Each for $479.82
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

101-00-8
199.06
IWKGJTDSJPLUCE-UHFFFAOYSA-N
225550
B12OC(CN(CC(O1)C)CC(O2)C)C
C9H18BNO3
MFCD03791298
triisopropanolamine cyclic borate, 3,7,10-trimethyl-2,8,9-trioxa-5-aza-1-borabicyclo 3.3.3 undecane, borester 21, triisopropanolamine borate, boric acid, tris 1-amino-2-propyl ester, 2-propanol, 1,1',1-nitrilotri-, cyclic borate, 3,7,10-trimethyl-4,6,11-trioxa-1-aza-5-borabicyclo 3.3.3 undecane, 2,8,9-trioxa-5-aza-1-borabicyclo 3.3.3 undecane, 3,7,10-trimethyl, triisopropanolamineborate
3,7,10-trimethyl-4,6,11-trioxa-1-aza-5-borabicyclo[3.3.3]undecane

Specifications

101-00-8
White to Yellow
98% min. (GC)
C9H18BNO3
100 g
IWKGJTDSJPLUCE-UHFFFAOYSA-N
3,7,10-trimethyl-4,6,11-trioxa-1-aza-5-borabicyclo[3.3.3]undecane
225550
98+%
Triisopropanolamine cyclic borate
150°C to 157°C
Authentic
Glass bottle
MFCD03791298
triisopropanolamine cyclic borate, 3,7,10-trimethyl-2,8,9-trioxa-5-aza-1-borabicyclo 3.3.3 undecane, borester 21, triisopropanolamine borate, boric acid, tris 1-amino-2-propyl ester, 2-propanol, 1,1',1-nitrilotri-, cyclic borate, 3,7,10-trimethyl-4,6,11-trioxa-1-aza-5-borabicyclo 3.3.3 undecane, 2,8,9-trioxa-5-aza-1-borabicyclo 3.3.3 undecane, 3,7,10-trimethyl, triisopropanolamineborate
B12OC(CN(CC(O1)C)CC(O2)C)C
199.06
199.06
Crystalline Chunks and Needles
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only