missing translation for 'onlineSavingsMsg'
Learn More

Triisopropanolamine cyclic borate, 98+%

Catalog No. AC269241000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC269241000 100 g Glass bottle
AC269245000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC269241000 Supplier Thermo Scientific Chemicals Supplier No. 269241000
Only null left

CAS: 101-00-8 | C9H18BNO3 | 199.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 101-00-8
Molecular Formula C9H18BNO3
Molecular Weight (g/mol) 199.06
MDL Number MFCD03791298
InChI Key IWKGJTDSJPLUCE-UHFFFAOYSA-N
Synonym triisopropanolamine cyclic borate, 3,7,10-trimethyl-2,8,9-trioxa-5-aza-1-borabicyclo 3.3.3 undecane, borester 21, triisopropanolamine borate, boric acid, tris 1-amino-2-propyl ester, 2-propanol, 1,1',1-nitrilotri-, cyclic borate, 3,7,10-trimethyl-4,6,11-trioxa-1-aza-5-borabicyclo 3.3.3 undecane, 2,8,9-trioxa-5-aza-1-borabicyclo 3.3.3 undecane, 3,7,10-trimethyl, triisopropanolamineborate
PubChem CID 225550
IUPAC Name 3,7,10-trimethyl-4,6,11-trioxa-1-aza-5-borabicyclo[3.3.3]undecane
SMILES B12OC(CN(CC(O1)C)CC(O2)C)C

Specifications

Melting Point 150°C to 157°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Quantity 100 g
Formula Weight 199.06
Percent Purity 98+%
Physical Form Crystalline Chunks and Needles
Chemical Name or Material Triisopropanolamine cyclic borate

RUO – Research Use Only