missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triheptylamine 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 155°C |
| Quantity | 25 mL |
| Formula Weight | 311.60 |
| Percent Purity | ≥98.0% (GC,T) |
| Grade | Not Available |
| Physical Form | Liquid |
| Chemical Name or Material | Triheptylamine [Reagent for Ion-Pair Chromatography] |
Safety and Handling
| TSCA | Yes |