CAS RN 2411-36-1
IUPAC Name:
triheptylamine
Synonyms:
triheptylamine
Molecular Weight (g/mol):
311.6
Molecular Formula:
C21H45N
InChi Key:
CLZGJKHEVKJLLS-UHFFFAOYSA-N
SMILES:
CCCCCCCN(CCCCCCC)CCCCCCC
1
–
1
of
1
results
1
Triheptylamine 98.0+%, TCI America™
CAS: 2411-36-1 Molecular Formula: C21H45N Molecular Weight (g/mol): 311.598 MDL Number: MFCD00041936 InChI Key: CLZGJKHEVKJLLS-UHFFFAOYSA-N Synonym: triheptylamine,tri-n-heptylamine,1-heptanamine, n,n-diheptyl,1-heptanamine,n,n-diheptyl,n,n-diheptyl-1-heptanamine #,clzgjkhevkjlls-uhfffaoysa PubChem CID: 75483 IUPAC Name: N,N-diheptylheptan-1-amine SMILES: CCCCCCCN(CCCCCCC)CCCCCCC