missing translation for 'onlineSavingsMsg'
Learn More

Trihalomethanes - WS, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111017031
Change view
Click to view available options
Quantity:
1.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017031 1.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017031 Supplier Merck Emd Millipore Supplier No. QC14561.2ML

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

Packaging Ampule
CAS 67-56-1
Flash Point 11°C - closed cup - Solvent
Type Certified Reference Material
Quantity 1.2 mL
Grade Certified Reference Material