missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trigonelline hydrochloride, MilliporeSigma™ Supelco™
Description
Featured Industries: Food and Beverages
Specifications
Specifications
| CAS | 6138-41-6 |
| Physical Form | Neat |
| Linear Formula | C7H7 NO2 · HCl |
| MDL Number | MFCD00077250 |
| Synonym | 1-Methylpyridinium-3-carboxylate hydrochloride; N-Methylnicotinic acid betaine hydrochloride |
| Type | Analytical Standard |
| Quantity | 1 g |
| Formula Weight | 173.60 |
| Grade | Analytical Standard |