missing translation for 'onlineSavingsMsg'
Learn More

Triglycidyl Isocyanurate 98.0+%, TCI America™

Catalog No. I042825G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I042825G 25 g
I0428100G 100 g
I0428500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I042825G Supplier TCI America Supplier No. I042825G

Chemical Identifiers

CAS 2451-62-9
Molecular Formula C12H15N3O6
Molecular Weight (g/mol) 297.267
MDL Number MFCD00080670
InChI Key OUPZKGBUJRBPGC-UHFFFAOYSA-N
Synonym Isocyanuric Acid Tris(2,3-epoxypropyl) Ester, Isocyanuric Acid Triglycidyl Ester, Tris(2,3-epoxypropyl) Isocyanurate
PubChem CID 17142
IUPAC Name 1,3,5-tris(oxiran-2-ylmethyl)-1,3,5-triazinane-2,4,6-trione
SMILES C1C(O1)CN2C(=O)N(C(=O)N(C2=O)CC3CO3)CC4CO4

Specifications

Melting Point 108°C
Color White
Quantity 25 g
UN Number 2811
Formula Weight 297.27
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Triglycidyl Isocyanurate
EINECSNumber (5)-1052
RTECSNumber XZ1994900
TSCA Yes