missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triflupromazine Hydrochloride 98.0+%, TCI America™
Supplier: TCI America T338925G
Specifications
| Triflupromazine Hydrochloride | |
| 177°C | |
| C18H20ClF3N2S | |
| 25 g | |
| triflupromazine hydrochloride, flumazin, fluorofen, neoprin, nivoman, vesprin hydrochloride, triflupromazine monohydrochloride, triflupromazine hcl, trifluopromazine hydrochloride | |
| [H+].[Cl-].CN(C)CCCN1C2=CC=CC=C2SC2=CC=C(C=C12)C(F)(F)F | |
| 388.88 | |
| CHEBI:9712 | |
| ≥98.0% (HPLC,N) |
| 1098-60-8 | |
| White-Yellow | |
| MFCD00058103 | |
| 2811 | |
| FTNWXGFYRHWUKG-UHFFFAOYSA-N | |
| hydrogen dimethyl({3-[2-(trifluoromethyl)-10H-phenothiazin-10-yl]propyl})amine chloride | |
| 66069 | |
| 388.88 | |
| Powder |
Chemical Identifiers
| 1098-60-8 | |
| 388.88 | |
| FTNWXGFYRHWUKG-UHFFFAOYSA-N | |
| 66069 | |
| hydrogen dimethyl({3-[2-(trifluoromethyl)-10H-phenothiazin-10-yl]propyl})amine chloride |
| C18H20ClF3N2S | |
| MFCD00058103 | |
| triflupromazine hydrochloride, flumazin, fluorofen, neoprin, nivoman, vesprin hydrochloride, triflupromazine monohydrochloride, triflupromazine hcl, trifluopromazine hydrochloride | |
| CHEBI:9712 | |
| [H+].[Cl-].CN(C)CCCN1C2=CC=CC=C2SC2=CC=C(C=C12)C(F)(F)F |
Safety and Handling
EINECSNumber : (9)-0927
RTECSNumber : SO8925000
TSCA : No