missing translation for 'onlineSavingsMsg'
Learn More

Triflupromazine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7137731
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33895G 5 g
T338925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T33895G Supplier TCI America Supplier No. T33895G

Chemical Identifiers

CAS 1098-60-8
Molecular Formula C18H20ClF3N2S
Molecular Weight (g/mol) 388.88
MDL Number MFCD00058103
InChI Key FTNWXGFYRHWUKG-UHFFFAOYSA-N
Synonym triflupromazine hydrochloride, flumazin, fluorofen, neoprin, nivoman, vesprin hydrochloride, triflupromazine monohydrochloride, triflupromazine hcl, trifluopromazine hydrochloride
PubChem CID 66069
ChEBI CHEBI:9712
IUPAC Name hydrogen dimethyl({3-[2-(trifluoromethyl)-10H-phenothiazin-10-yl]propyl})amine chloride
SMILES [H+].[Cl-].CN(C)CCCN1C2=CC=CC=C2SC2=CC=C(C=C12)C(F)(F)F

Specifications

Melting Point 177°C
Color White-Yellow
Quantity 5 g
UN Number 2811
Formula Weight 388.88
Percent Purity ≥98.0% (HPLC,N)
Physical Form Powder
Chemical Name or Material Triflupromazine Hydrochloride
EINECSNumber (9)-0927
RTECSNumber SO8925000
TSCA No