missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Trifluoromethyl)tris(triphenylphosphine)copper(I) 98.0+%, TCI America™
$689.70 - $2356.16
Chemical Identifiers
| CAS | 325810-07-9 |
|---|---|
| Molecular Formula | C55H45CuF3P3 |
| Molecular Weight (g/mol) | 919.427 |
| InChI Key | ADEZRALRKPUAPZ-UHFFFAOYSA-N |
| Synonym | Tris(triphenylphosphine)(trifluoromethyl)copper(I) |
| PubChem CID | 85433410 |
| IUPAC Name | copper(1+);trifluoromethane;triphenylphosphane |
| SMILES | C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[C-](F)(F)F.[Cu+] |
Chemical Identifiers
| 325810-07-9 | |
| 919.427 | |
| Tris(triphenylphosphine)(trifluoromethyl)copper(I) | |
| copper(1+);trifluoromethane;triphenylphosphane |
| C55H45CuF3P3 | |
| ADEZRALRKPUAPZ-UHFFFAOYSA-N | |
| 85433410 | |
| C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[C-](F)(F)F.[Cu+] |
Specifications
| 325810-07-9 | |
| White | |
| 1 g | |
| ADEZRALRKPUAPZ-UHFFFAOYSA-N | |
| copper(1+);trifluoromethane;triphenylphosphane | |
| 85433410 | |
| ≥98.0% (T) | |
| (Trifluoromethyl)tris(triphenylphosphine)copper(I) |
| 155°C | |
| C55H45CuF3P3 | |
| Tris(triphenylphosphine)(trifluoromethyl)copper(I) | |
| C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[C-](F)(F)F.[Cu+] | |
| 919.427 | |
| 919.43 | |
| Crystalline Powder |
Safety and Handling
TSCA : No