missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Trifluoromethyl)tris(triphenylphosphine)copper(I) 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 155°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 919.43 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | (Trifluoromethyl)tris(triphenylphosphine)copper(I) |
Safety and Handling
| TSCA | No |