missing translation for 'onlineSavingsMsg'
Learn More

(Trifluoromethyl)tris(triphenylphosphine)copper(I) 98.0+%, TCI America™

Catalog No. T28831G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T28831G 1 g
T28835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T28831G Supplier TCI America Supplier No. T28831G
Only null left

Chemical Identifiers

CAS 325810-07-9
Molecular Formula C55H45CuF3P3
Molecular Weight (g/mol) 919.427
InChI Key ADEZRALRKPUAPZ-UHFFFAOYSA-N
Synonym Tris(triphenylphosphine)(trifluoromethyl)copper(I)
PubChem CID 85433410
IUPAC Name copper(1+);trifluoromethane;triphenylphosphane
SMILES C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[C-](F)(F)F.[Cu+]

Specifications

Melting Point 155°C
Color White
Quantity 1 g
Formula Weight 919.43
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (Trifluoromethyl)tris(triphenylphosphine)copper(I)