missing translation for 'onlineSavingsMsg'
Learn More

(Trifluoromethyl)cyclohexane 98.0+%, TCI America™

$83.89 - $231.14

Chemical Identifiers

CAS 401-75-2
Molecular Formula C7H11F3
Molecular Weight (g/mol) 152.16
MDL Number MFCD03412289
InChI Key QXPZOKVSFMRGMQ-UHFFFAOYSA-N
PubChem CID 6432413
IUPAC Name trifluoromethylcyclohexane
SMILES C1CCC(CC1)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T36285G
View Documents
TCI America
T36285G
5 g N/A
Only null left
 
T362825G
View Documents
TCI America
T362825G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

401-75-2
152.16
QXPZOKVSFMRGMQ-UHFFFAOYSA-N
trifluoromethylcyclohexane
C7H11F3
MFCD03412289
6432413
C1CCC(CC1)C(F)(F)F

Specifications

401-75-2
107°C
MFCD03412289
1993
C1CCC(CC1)C(F)(F)F
152.16
152.16
Liquid
Colorless
C7H11F3
5 g
QXPZOKVSFMRGMQ-UHFFFAOYSA-N
trifluoromethylcyclohexane
6432413
≥98.0% (GC)
(Trifluoromethyl)cyclohexane
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'