accessibility menu, dialog, popup

UserName

FFF

CAS RN 401-75-2

IUPAC Name: (trifluoromethyl)cyclohexane
Synonyms: perfluoromethylcyclohexane
Molecular Weight (g/mol): 152.16
Molecular Formula: C7H11F3
InChi Key: QXPZOKVSFMRGMQ-UHFFFAOYSA-N
SMILES: FC(F)(F)C1CCCCC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results