missing translation for 'onlineSavingsMsg'
Learn More

(Trifluoromethyl)cyclohexane 98.0+%, TCI America™

Catalog No. T36285G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T36285G 5 g
T362825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T36285G Supplier TCI America Supplier No. T36285G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 401-75-2
Molecular Formula C7H11F3
Molecular Weight (g/mol) 152.16
MDL Number MFCD03412289
InChI Key QXPZOKVSFMRGMQ-UHFFFAOYSA-N
PubChem CID 6432413
IUPAC Name trifluoromethylcyclohexane
SMILES C1CCC(CC1)C(F)(F)F

Specifications

Color Colorless
Boiling Point 107°C
Quantity 5 g
UN Number 1993
Formula Weight 152.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (Trifluoromethyl)cyclohexane