missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trifluoromethanesulfonic Acid 98.0+%, TCI America™
Supplier: TCI America T075125G
Specifications
| Trifluoromethanesulfonic Acid | |
| Yellow | |
| CHF3O3S | |
| 25 g | |
| triflic acid, perfluoromethanesulfonic acid, trifluoromethanesulphonic acid, tfmsa, trifluoromethane sulfonic acid, tfoh, methanesulfonic acid, trifluoro, hotf, trifluoromethylsulfonic acid, trimsylate | |
| OS(=O)(=O)C(F)(F)F | |
| 150.07 | |
| CHEBI:48511 | |
| ≥98.0% (T) |
| 1493-13-6 | |
| 162°C | |
| MFCD00007514 | |
| 3265 | |
| ITMCEJHCFYSIIV-UHFFFAOYSA-N | |
| trifluoromethanesulfonic acid | |
| 62406 | |
| 150.07 | |
| Liquid |
Chemical Identifiers
| 1493-13-6 | |
| 150.07 | |
| ITMCEJHCFYSIIV-UHFFFAOYSA-N | |
| 62406 | |
| trifluoromethanesulfonic acid |
| CHF3O3S | |
| MFCD00007514 | |
| triflic acid, perfluoromethanesulfonic acid, trifluoromethanesulphonic acid, tfmsa, trifluoromethane sulfonic acid, tfoh, methanesulfonic acid, trifluoro, hotf, trifluoromethylsulfonic acid, trimsylate | |
| CHEBI:48511 | |
| OS(=O)(=O)C(F)(F)F |
Safety and Handling
EINECSNumber : (2)-2809
TSCA : Yes