accessibility menu, dialog, popup

UserName

OHSOOFFF

CAS RN 1493-13-6

IUPAC Name: trifluoromethanesulfonic acid
Synonyms: trifluoromethanesulfonic acid|triflic acid|Cu(OTf)2|HOTf|In(OTf)3|TfOH|acide triflique|AgOTf cpd|tfmsa cpd|trifluormethansulfonsaeure
Molecular Weight (g/mol): 150.07
Molecular Formula: CHF3O3S
InChi Key: ITMCEJHCFYSIIV-UHFFFAOYSA-N
SMILES: OS(=O)(=O)C(F)(F)F
Boiling Point 
  • (3)
  • (5)
Percent Purity 
  • (3)
  • (5)
Physical Form 
  • (5)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (3)
  • (5)
  • (3)
  • (5)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)