missing translation for 'onlineSavingsMsg'
Learn More

Trifluoromethanesulfonic Acid 98.0+%, TCI America™

Catalog No. T075110G
Change view
Click to view available options
Quantity:
10 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T075110G 10 g
T075125G 25 g
T0751250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T075110G Supplier TCI America Supplier No. T075110G

Chemical Identifiers

CAS 1493-13-6
Molecular Formula CHF3O3S
Molecular Weight (g/mol) 150.07
MDL Number MFCD00007514
InChI Key ITMCEJHCFYSIIV-UHFFFAOYSA-N
Synonym triflic acid, perfluoromethanesulfonic acid, trifluoromethanesulphonic acid, tfmsa, trifluoromethane sulfonic acid, tfoh, methanesulfonic acid, trifluoro, hotf, trifluoromethylsulfonic acid, trimsylate
PubChem CID 62406
ChEBI CHEBI:48511
IUPAC Name trifluoromethanesulfonic acid
SMILES OS(=O)(=O)C(F)(F)F

Specifications

Color Yellow
Boiling Point 162°C
Quantity 10 g
UN Number 3265
Formula Weight 150.07
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Trifluoromethanesulfonic Acid
EINECSNumber (2)-2809
TSCA Yes