Learn More
Trifluoroiodomethane, 97+%
Description
Trifluoroiodomethane is used as a gaseous fire suppression flooding agent for in-flight aircraft and electronic equipment fires. It is also an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. It is involved in the rhodium-catalyzed alfa-trifluoromethylation of alfa,beta-unsaturatedketones. It plays an important role as catalyst in the enantioselective alfa -trifluoromethylation of aldehydes through photoredox organocatalysis using a readily available iridium photocatalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 2.361 g/mL |
| Boiling Point | -22.5°C |
| Flash Point | None |
| Packaging | Cylinder |
| Refractive Index | 1.379 |
| Quantity | 25 g |
| UN Number | UN1956 |
| Beilstein | 1732740 |
| Sensitivity | Light sensitive |
| Solubility Information | Slightly soluble in water. |
| Show More |
Safety and Handling
| Hazard Category | H341 |
| Hazard Statement | GHS H Statement H280-H341 Contains gas under pressure may explode if heated. Suspected of causing genetic defects. |
| Precautionary Statement | P201-P202-P281-P308+P313-P501c |
| DOTInformation | Transport Hazard Class: 2.2; Proper Shipping Name: COMPRESSED GAS N.O.S. |
| EINECSNumber | 219-014-5 |
| RTECSNumber | PB6975000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only