missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroiodomethane, 97+%

Catalog No. AA4147914
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4147914 25 g
AA4147922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4147914 Supplier Thermo Scientific Chemicals Supplier No. 04147914

CAS: 2314-97-8 | CF3I | 195.911 g/mol

Trifluoroiodomethane is used as a gaseous fire suppression flooding agent for in-flight aircraft and electronic equipment fires. It is also an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. It is involved in the rhodium-catalyzed alfa-trifluoromethylation of alfa,beta-unsaturatedketones. It plays an important role as catalyst in the enantioselective alfa -trifluoromethylation of aldehydes through photoredox organocatalysis using a readily available iridium photocatalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2314-97-8
Molecular Formula CF3I
Molecular Weight (g/mol) 195.911
MDL Number MFCD00001060
InChI Key VPAYJEUHKVESSD-UHFFFAOYSA-N
Synonym trifluoromethyl iodide, iodotrifluoromethane, methane, trifluoroiodo, perfluoromethyl iodide, trifluoro iodo methane, trifluoromethyliodide, monoiodotrifluoromethane, freon 13t1, unii-42a379kb0u, ccris 7782
PubChem CID 16843
IUPAC Name trifluoro(iodo)methane
SMILES C(F)(F)(F)I

Specifications

Density 2.361 g/mL
Boiling Point -22.5°C
Flash Point None
Packaging Cylinder
Refractive Index 1.379
Quantity 25 g
UN Number UN1956
Beilstein 1732740
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 195.91
Percent Purity ≥97%
Chemical Name or Material Trifluoroiodomethane
Show More Show Less
Hazard Category H341
Hazard Statement GHS H Statement
H280-H341
Contains gas under pressure
may explode if heated.
Suspected of causing genetic defects.
Precautionary Statement P201-P202-P281-P308+P313-P501c
DOTInformation Transport Hazard Class: 2.2; Proper Shipping Name: COMPRESSED GAS N.O.S.
EINECSNumber 219-014-5
RTECSNumber PB6975000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only