missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetylacetone 98.0+%, TCI America™

Catalog No. p-7136281
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T043425G 25 g
T0434100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T043425G Supplier TCI America Supplier No. T043425G

Chemical Identifiers

CAS 367-57-7
Molecular Formula C5H5F3O2
Molecular Weight (g/mol) 154.09
MDL Number MFCD00000427
InChI Key SHXHPUAKLCCLDV-UHFFFAOYSA-N
Synonym 1,1,1-trifluoro-2,4-pentanedione, trifluoroacetylacetone, 1,1,1-trifluoroacetylacetone, trifluoroacetyl acetone, acetyl trifluoroacetone, 2,4-pentanedione, 1,1,1-trifluoro, tfpd-h, unii-9n20a8g8sw, alpha,alpha,alpha-trifluoroacetylacetone, a,a,a-trifluoroacetylacetone
PubChem CID 73943
IUPAC Name 1,1,1-trifluoropentane-2,4-dione
SMILES CC(=O)CC(=O)C(F)(F)F

Specifications

Color Yellow
Boiling Point 101°C
Quantity 25 g
UN Number 1224
Formula Weight 154.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Trifluoroacetylacetone