accessibility menu, dialog, popup

UserName

H3COOFFF

CAS RN 367-57-7

IUPAC Name: 1,1,1-trifluoropentane-2,4-dione
Synonyms: 1,1,1-trifluoropentane-2,4-dione
Molecular Weight (g/mol): 154.09
Molecular Formula: C5H5F3O2
InChi Key: SHXHPUAKLCCLDV-UHFFFAOYSA-N
SMILES: CC(=O)CC(=O)C(F)(F)F
Boiling Point 
  • (2)
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (3)
  • (1)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)