missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trifluoroacetic acid, Puriss. p.a., for HPLC, ≥99.0% (GC), Honeywell Fluka™
Puriss. p.a., for HPLC,≥99.0% (GC)
$1739.65 - $1739.65
Chemical Identifiers
| CAS | 76-05-1 |
|---|---|
| Molecular Formula | C2HF3O2 |
| Molecular Weight (g/mol) | 114.02 |
| MDL Number | MFCD00004169 |
| InChI Key | DTQVDTLACAAQTR-UHFFFAOYSA-N |
| Synonym | trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova |
| PubChem CID | 6422 |
| ChEBI | CHEBI:45892 |
| SMILES | OC(=O)C(F)(F)F |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 76-05-1 | |
| 114.02 | |
| DTQVDTLACAAQTR-UHFFFAOYSA-N | |
| 6422 | |
| OC(=O)C(F)(F)F |
| C2HF3O2 | |
| MFCD00004169 | |
| trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova | |
| CHEBI:45892 |
Specifications
| 76-05-1 | |
| CF3COOH | |
| UN2699 | |
| trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova | |
| OC(=O)C(F)(F)F | |
| 114.02 | |
| CHEBI:45892 | |
| ≥99% (GC) | |
| 250 mL | |
| 3.9 (vs air) | |
| Trifluoroacetic acid |
| C2HF3O2 | |
| MFCD00004169 | |
| 742035 | |
| DTQVDTLACAAQTR-UHFFFAOYSA-N | |
| trifluoroacetic acid | |
| 6422 | |
| 114.02g/mol | |
| 1.489g/mL at 20°C | |
| >100°C (>212°F) | |
| 97.5 mmHg (20°C) |
Safety and Handling
P273-P280-P305 + P351 + P338-P310
RTECSNumber : AJ9625000