missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic acid, Puriss. p.a., for HPLC, ≥99.0% (GC), Honeywell Fluka™

Catalog No. 6001754
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001754 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6001754 Supplier Honeywell Chemicals Supplier No. 91707250ML
Only null left

Puriss. p.a., for HPLC,≥99.0% (GC)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 76-05-1
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 114.02
MDL Number MFCD00004169
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
Synonym trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
PubChem CID 6422
ChEBI CHEBI:45892
SMILES OC(=O)C(F)(F)F

Specifications

Linear Formula CF3COOH
UN Number UN2699
Beilstein 742035
IUPAC Name trifluoroacetic acid
Formula Weight 114.02g/mol
Percent Purity ≥99% (GC)
Density 1.489g/mL at 20°C
Quantity 250 mL
Flash Point >100°C (>212°F)
Vapor Density 3.9 (vs air)
Vapor Pressure 97.5 mmHg (20°C)
Chemical Name or Material Trifluoroacetic acid
Show More Show Less
Hazard Statement P273-P280-P305 + P351 + P338-P310
RTECSNumber AJ9625000