missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trifluoroacetamidine, tech. 85%
Description
Trifluoroacetamidine is used as probe for determination of membrane potential and extra/intracellular volume of erythrocytes by fluorine-19 NMR studies. It is a reagent used as protected ammonia equivalent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Trifluoroacetamidine is used as probe for determination of membrane potential and extra/intracellular volume of erythrocytes by fluorine-19 NMR studies. It is a reagent used as protected ammonia equivalent.
Solubility
Fully miscible in water.
Notes
Store at -20°C. Incompatible with oxidizing agents. Moisture Sensitive.
Trifluoroacetamidine is used as probe for determination of membrane potential and extra/intracellular volume of erythrocytes by fluorine-19 NMR studies. It is a reagent used as protected ammonia equivalent.
Solubility
Fully miscible in water.
Notes
Store at -20°C. Incompatible with oxidizing agents. Moisture Sensitive.
Specifications
Specifications
| Density | 1.45 |
| Boiling Point | 35°C to 37°C (11 mmHg) |
| Refractive Index | 1.382 |
| Quantity | 5 g |
| UN Number | UN3267 |
| Beilstein | 605978 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Fully miscible in water. |
| Formula Weight | 112.05 |
| Percent Purity | 85% |
| Show More |
Safety and Handling
| Hazard Category | H314-H335 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Hazard Class: Y; Packaging Group: II |
| EINECSNumber | 000-000-0 |
| TSCA | No |
| Recommended Storage | Store at -20°C |
RUO – Research Use Only