missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetamide 98.0+%, TCI America™

Catalog No. p-7136354
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T059825G 25 g
T0598100G 100 g
T0598500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T059825G Supplier TCI America Supplier No. T059825G
Only null left

Chemical Identifiers

CAS 354-38-1
Molecular Formula C2H2F3NO
Molecular Weight (g/mol) 113.04
MDL Number MFCD00008008
InChI Key NRKYWOKHZRQRJR-UHFFFAOYSA-N
Synonym trifluoroacetamide, acetamide, 2,2,2-trifluoro, unii-36mu16byfk, n-trifluoroacetamide, 2,2,2-trifluoro-acetamide, 36mu16byfk, trifluroacetamide, trifluoro-acetamide, snylgqhjptadtaetp@, pubchem12622
PubChem CID 67717
IUPAC Name trifluoroacetamide
SMILES NC(=O)C(F)(F)F

Specifications

Melting Point 72°C
Color White
Boiling Point 163°C
Quantity 25 g
Formula Weight 113.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Trifluoroacetamide
EINECSNumber (2)-3531
TSCA Yes