missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha,alpha-Trifluoro-m-cresol, 98+%

Catalog No. AC168470250
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC168470250 25 mL Glass bottle
AC168471000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC168470250 Supplier Thermo Scientific Chemicals Supplier No. 168470250

CAS: 98-17-9 | C7H5F3O | 162.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 98-17-9
Molecular Formula C7H5F3O
Molecular Weight (g/mol) 162.11
MDL Number MFCD00002299
InChI Key UGEJOEBBMPOJMT-UHFFFAOYSA-N
Synonym 3-trifluoromethyl phenol, 3-hydroxybenzotrifluoride, m-trifluoromethylphenol, m-hydroxybenzotrifluoride, phenol, 3-trifluoromethyl, m-trifluoromethyl phenol, alpha,alpha,alpha-trifluoro-m-cresol, a,a,a-trifluoro-m-cresol
PubChem CID 7376
IUPAC Name 3-(trifluoromethyl)phenol
SMILES C1=CC(=CC(=C1)O)C(F)(F)F

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -1.8°C
Color Yellow
Density 1.3300g/mL
Boiling Point 178°C to 179°C
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Refractive Index 1.4570 to 1.459
Linear Formula CF3C6H4OH
Quantity 25 mL
Beilstein 06, I, 187
Specific Gravity 1.33
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most organic solvents
Formula Weight 162.11
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material α, α, α-Trifluoro-m-cresol, 98+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 202-645-5
RTECSNumber GP3510000
TSCA TSCA

RUO – Research Use Only