missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trifluoperazine Dihydrochloride 98.0+%, TCI America™
Supplier: TCI America T284925G
Specifications
| Trifluoperazine Dihydrochloride | |
| 293°C | |
| C21H26Cl2F3N3S | |
| 25 g | |
| BXDAOUXDMHXPDI-UHFFFAOYSA-N | |
| 10-[3-(4-methylpiperazin-1-yl)propyl]-2-(trifluoromethyl)phenothiazine;dihydrochloride | |
| 66064 | |
| 480.42 | |
| Crystalline Powder |
| 440-17-5 | |
| White-Yellow | |
| MFCD00012656 | |
| trifluoperazine dihydrochloride, trifluoperazine hydrochloride, terfluzine, stelazine, triphthazine, eskazinyl, jatroneural, tryptazine, trazine, fluoperazine | |
| CN1CCN(CC1)CCCN2C3=CC=CC=C3SC4=C2C=C(C=C4)C(F)(F)F.Cl.Cl | |
| 480.415 | |
| CHEBI:9710 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 440-17-5 | |
| 480.415 | |
| BXDAOUXDMHXPDI-UHFFFAOYSA-N | |
| 66064 | |
| 10-[3-(4-methylpiperazin-1-yl)propyl]-2-(trifluoromethyl)phenothiazine;dihydrochloride |
| C21H26Cl2F3N3S | |
| MFCD00012656 | |
| trifluoperazine dihydrochloride, trifluoperazine hydrochloride, terfluzine, stelazine, triphthazine, eskazinyl, jatroneural, tryptazine, trazine, fluoperazine | |
| CHEBI:9710 | |
| CN1CCN(CC1)CCCN2C3=CC=CC=C3SC4=C2C=C(C=C4)C(F)(F)F.Cl.Cl |
Safety and Handling
RTECSNumber : SP1750000
TSCA : Yes
Recommended Storage : Freezer