missing translation for 'onlineSavingsMsg'
Learn More

Trifluoperazine Dihydrochloride 98.0+%, TCI America™

$186.41 - $646.83

Chemical Identifiers

CAS 440-17-5
Molecular Formula C21H26Cl2F3N3S
Molecular Weight (g/mol) 480.415
MDL Number MFCD00012656
InChI Key BXDAOUXDMHXPDI-UHFFFAOYSA-N
Synonym trifluoperazine dihydrochloride, trifluoperazine hydrochloride, terfluzine, stelazine, triphthazine, eskazinyl, jatroneural, tryptazine, trazine, fluoperazine
PubChem CID 66064
ChEBI CHEBI:9710
IUPAC Name 10-[3-(4-methylpiperazin-1-yl)propyl]-2-(trifluoromethyl)phenothiazine;dihydrochloride
SMILES CN1CCN(CC1)CCCN2C3=CC=CC=C3SC4=C2C=C(C=C4)C(F)(F)F.Cl.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T28495G
View Documents
TCI America
T28495G
5 g N/A
Only null left
 
T284925G
View Documents
TCI America
T284925G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

440-17-5
480.415
BXDAOUXDMHXPDI-UHFFFAOYSA-N
66064
10-[3-(4-methylpiperazin-1-yl)propyl]-2-(trifluoromethyl)phenothiazine;dihydrochloride
C21H26Cl2F3N3S
MFCD00012656
trifluoperazine dihydrochloride, trifluoperazine hydrochloride, terfluzine, stelazine, triphthazine, eskazinyl, jatroneural, tryptazine, trazine, fluoperazine
CHEBI:9710
CN1CCN(CC1)CCCN2C3=CC=CC=C3SC4=C2C=C(C=C4)C(F)(F)F.Cl.Cl

Specifications

440-17-5
White-Yellow
MFCD00012656
trifluoperazine dihydrochloride, trifluoperazine hydrochloride, terfluzine, stelazine, triphthazine, eskazinyl, jatroneural, tryptazine, trazine, fluoperazine
CN1CCN(CC1)CCCN2C3=CC=CC=C3SC4=C2C=C(C=C4)C(F)(F)F.Cl.Cl
480.415
CHEBI:9710
≥98.0% (HPLC,T)
Trifluoperazine Dihydrochloride
293°C
C21H26Cl2F3N3S
5 g
BXDAOUXDMHXPDI-UHFFFAOYSA-N
10-[3-(4-methylpiperazin-1-yl)propyl]-2-(trifluoromethyl)phenothiazine;dihydrochloride
66064
480.42
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

RTECSNumber : SP1750000

TSCA : Yes

Recommended Storage : Freezer

SDS
missing translation for 'documents'

missing translation for 'documents'