missing translation for 'onlineSavingsMsg'
Learn More

Triethylvinylsilane 96.0+%, TCI America™

Supplier:  TCI America T170325ML

 View more versions of this product

Catalog No. T170325ML

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Triethylvinylsilane
Yellow
C8H18Si
25 mL
vinyltriethylsilane, triethylvinylsilane, triethyl vinyl silane, vinyl triethyl silane, ethenyl-triethylsilane, triethylsilyl ethylene, ethenyl triethyl silane, pubchem13130, triethyl vinyl silane #
CC[Si](CC)(CC)C=C
142.32
142.32
Liquid
1112-54-5
147°C
MFCD00041345
1993
HBWGDHDXAMFADB-UHFFFAOYSA-N
ethenyltriethylsilane
559195
≥96.0% (GC)

Chemical Identifiers

1112-54-5
142.32
HBWGDHDXAMFADB-UHFFFAOYSA-N
559195
CC[Si](CC)(CC)C=C
C8H18Si
MFCD00041345
vinyltriethylsilane, triethylvinylsilane, triethyl vinyl silane, vinyl triethyl silane, ethenyl-triethylsilane, triethylsilyl ethylene, ethenyl triethyl silane, pubchem13130, triethyl vinyl silane #
ethenyltriethylsilane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions