missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethylvinylsilane 96.0+%, TCI America™
$123.03 - $480.92
Chemical Identifiers
| CAS | 1112-54-5 |
|---|---|
| Molecular Formula | C8H18Si |
| Molecular Weight (g/mol) | 142.32 |
| MDL Number | MFCD00041345 |
| InChI Key | HBWGDHDXAMFADB-UHFFFAOYSA-N |
| Synonym | vinyltriethylsilane, triethylvinylsilane, triethyl vinyl silane, vinyl triethyl silane, ethenyl-triethylsilane, triethylsilyl ethylene, ethenyl triethyl silane, pubchem13130, triethyl vinyl silane # |
| PubChem CID | 559195 |
| IUPAC Name | ethenyltriethylsilane |
| SMILES | CC[Si](CC)(CC)C=C |
Chemical Identifiers
| 1112-54-5 | |
| 142.32 | |
| HBWGDHDXAMFADB-UHFFFAOYSA-N | |
| 559195 | |
| CC[Si](CC)(CC)C=C |
| C8H18Si | |
| MFCD00041345 | |
| vinyltriethylsilane, triethylvinylsilane, triethyl vinyl silane, vinyl triethyl silane, ethenyl-triethylsilane, triethylsilyl ethylene, ethenyl triethyl silane, pubchem13130, triethyl vinyl silane # | |
| ethenyltriethylsilane |
Specifications
| 1112-54-5 | |
| 147°C | |
| MFCD00041345 | |
| 1993 | |
| HBWGDHDXAMFADB-UHFFFAOYSA-N | |
| ethenyltriethylsilane | |
| 559195 | |
| ≥96.0% (GC) | |
| Triethylvinylsilane |
| Yellow | |
| C8H18Si | |
| 5 mL | |
| vinyltriethylsilane, triethylvinylsilane, triethyl vinyl silane, vinyl triethyl silane, ethenyl-triethylsilane, triethylsilyl ethylene, ethenyl triethyl silane, pubchem13130, triethyl vinyl silane # | |
| CC[Si](CC)(CC)C=C | |
| 142.32 | |
| 142.32 | |
| Liquid |
Safety and Handling
TSCA : No