missing translation for 'onlineSavingsMsg'
Learn More

Triethylsulfonium bis(trifluoromethylsulfonyl)imide, 98%

Catalog No. p-7029630
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2711606 5 g
AAH2711618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2711606 Supplier Thermo Scientific Chemicals Supplier No. H2711606
Only null left

CAS: 321746-49-0 | C8H15F6NO4S3 | 399.381 g/mol

Proton-Conducting Properties of a Bronsted Acid-Base Ionic Liquid and Ionic Melts Consisting of Bis(trifluoromethanesulfonyl)imide and Benzimidazole for Fuel Cell Electrolytes. Symmetric and aliphatic trialkylsulfonium cation form room temperature molten salts (RTMS) with bis(trifluoromethylsulfonyl)imide (TFSI).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Proton-Conducting Properties of a Bronsted Acid-Base Ionic Liquid and Ionic Melts Consisting of Bis(trifluoromethanesulfonyl)imide and Benzimidazole for Fuel Cell Electrolytes. Symmetric and aliphatic trialkylsulfonium cation form room temperature molten salts (RTMS) with bis(trifluoromethylsulfonyl)imide (TFSI).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 321746-49-0
Molecular Formula C8H15F6NO4S3
Molecular Weight (g/mol) 399.381
MDL Number MFCD08276438
InChI Key BLODSRKENWXTLO-UHFFFAOYSA-N
Synonym triethylsulfonium bis trifluoromethylsulfonyl imide, triethylsulfonium bis trifluoromethanesulfonyl imide, set3 ntf2, dsstox_cid_27864, dsstox_rid_82617, dsstox_gsid_47888, bistriflylimide anion; triethylsulfanium, triethylsulfonium bis trifluoromethylsulfonyl imide, for electrochemistry qnmr
PubChem CID 16211135
IUPAC Name bis(trifluoromethylsulfonyl)azanide;triethylsulfanium
SMILES CC[S+](CC)CC.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Specifications

Quantity 5 g
Formula Weight 399.39
Percent Purity 98%
Chemical Name or Material Triethylsulfonium bis(trifluoromethylsulfonyl)imide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only