accessibility menu, dialog, popup

UserName

CAS RN 321746-49-0

IUPAC Name: triethylsulfanium; trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane
Synonyms: bistriflylimide anion; triethylsulfanium
Molecular Weight (g/mol): 399.38
Molecular Formula: C8H15F6NO4S3
InChi Key: BLODSRKENWXTLO-UHFFFAOYSA-N
SMILES: CC[S+](CC)CC.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)