missing translation for 'onlineSavingsMsg'
Learn More

Triethylsulfonium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™

Catalog No. T23145G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T23145G Supplier TCI America Supplier No. T23145G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 321746-49-0
Molecular Formula C8H15F6NO4S3
Molecular Weight (g/mol) 399.381
InChI Key BLODSRKENWXTLO-UHFFFAOYSA-N
Synonym triethylsulfonium bis trifluoromethylsulfonyl imide, triethylsulfonium bis trifluoromethanesulfonyl imide, set3 ntf2, dsstox_cid_27864, dsstox_rid_82617, dsstox_gsid_47888, bistriflylimide anion; triethylsulfanium, triethylsulfonium bis trifluoromethylsulfonyl imide, for electrochemistry qnmr
PubChem CID 16211135
IUPAC Name bis(trifluoromethylsulfonyl)azanide;triethylsulfanium
SMILES CC[S+](CC)CC.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Specifications

Color Yellow
Quantity 5 g
Formula Weight 399.38
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Triethylsulfonium Bis(trifluoromethanesulfonyl)imide