missing translation for 'onlineSavingsMsg'
Learn More

Triethylsilylacetylene 96.0+%, TCI America™

Catalog No. T23871G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23871G 1 g
T23875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T23871G Supplier TCI America Supplier No. T23871G

Chemical Identifiers

CAS 1777-03-3
Molecular Formula C8H16Si
Molecular Weight (g/mol) 140.30
MDL Number MFCD00075373
InChI Key FWSPXZXVNVQHIF-UHFFFAOYSA-N
Synonym triethylsilyl acetylene, ethynyltriethylsilane, triethyl ethynyl silane, triethylsilylacetylene, silane, triethylethynyl, triethylsilyl acetylen, triethylsilyl-acetylene, triethyl-ethynyl-silane
PubChem CID 637947
IUPAC Name triethyl(ethynyl)silane
SMILES CC[Si](CC)(CC)C#C

Specifications

Color Yellow
Boiling Point 136°C
Quantity 1 g
UN Number 1993
Formula Weight 140.30
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethylsilylacetylene