missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethylsilane 98.0+%, TCI America™
Supplier: TCI America T066225ML
Specifications
| 108°C | |
| Colorless | |
| Liquid | |
| C6H15Si | |
| 1993 | |
| QXTIBZLKQPJVII-UHFFFAOYSA-N | |
| triethylsilicon | |
| 6327258 | |
| ≥98.0% (GC) |
| Triethylsilane | |
| 617-86-7 | |
| 25 mL | |
| MFCD00009018 | |
| triethylsilane, hydride, silane, triethyl, triethylhydrosilane, triethylsilyl, silane e3h, c6h16si, triethylsilyl radical, silane, triethyl-,, triethylsilane tes | |
| CC[Si](CC)CC | |
| 115.271 | |
| 116.28 |
Chemical Identifiers
| 617-86-7 | |
| 115.271 | |
| QXTIBZLKQPJVII-UHFFFAOYSA-N | |
| 6327258 | |
| CC[Si](CC)CC |
| C6H15Si | |
| MFCD00009018 | |
| triethylsilane, hydride, silane, triethyl, triethylhydrosilane, triethylsilyl, silane e3h, c6h16si, triethylsilyl radical, silane, triethyl-,, triethylsilane tes | |
| triethylsilicon |
Safety and Handling
TSCA : Yes