missing translation for 'onlineSavingsMsg'
Learn More

Triethylsilane 98.0+%, TCI America™

$86.64 - $428.73

Chemical Identifiers

CAS 617-86-7
Molecular Formula C6H15Si
Molecular Weight (g/mol) 115.271
MDL Number MFCD00009018
InChI Key QXTIBZLKQPJVII-UHFFFAOYSA-N
Synonym triethylsilane, hydride, silane, triethyl, triethylhydrosilane, triethylsilyl, silane e3h, c6h16si, triethylsilyl radical, silane, triethyl-,, triethylsilane tes
PubChem CID 6327258
IUPAC Name triethylsilicon
SMILES CC[Si](CC)CC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T066225ML
View Documents
TCI America
T066225ML
25 mL
Each for $86.64
Only null left
 
T0662250ML
View Documents
TCI America
T0662250ML
250 mL
Each for $428.73
Only null left
 
Description

Description

Specifications

Chemical Identifiers

617-86-7
115.271
QXTIBZLKQPJVII-UHFFFAOYSA-N
6327258
CC[Si](CC)CC
C6H15Si
MFCD00009018
triethylsilane, hydride, silane, triethyl, triethylhydrosilane, triethylsilyl, silane e3h, c6h16si, triethylsilyl radical, silane, triethyl-,, triethylsilane tes
triethylsilicon

Specifications

108°C
617-86-7
25 mL
MFCD00009018
triethylsilane, hydride, silane, triethyl, triethylhydrosilane, triethylsilyl, silane e3h, c6h16si, triethylsilyl radical, silane, triethyl-,, triethylsilane tes
CC[Si](CC)CC
115.271
116.28
Triethylsilane
Colorless
Liquid
C6H15Si
1993
QXTIBZLKQPJVII-UHFFFAOYSA-N
triethylsilicon
6327258
≥98.0% (GC)
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents