missing translation for 'onlineSavingsMsg'
Learn More

Triethylphenylammonium Iodide 98.0+%, TCI America™

Catalog No. p024225g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P024225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P024225G Supplier TCI America Supplier No. P024225G
Only null left

Chemical Identifiers

CAS 1010-19-1
Molecular Formula C12H20IN
Molecular Weight (g/mol) 305.203
MDL Number MFCD00050224
InChI Key WMSWXWGJYOIACA-UHFFFAOYSA-M
Synonym triethylphenylammonium iodide, n,n,n-triethylbenzenaminium iodide, n,n,n-triethylanilinium iodide, phenyltriethylammonium iodide, benzenaminium, n,n,n-triethyl-, iodide, benzenaminium, n,n,n-triethyl-, iodide 1:1, ammonium, triethylphenyl-, iodide, acmc-1bzum, phenyltriathylammoniumjodid, triethylphenylammoniumiodide
PubChem CID 70524
IUPAC Name triethyl(phenyl)azanium;iodide
SMILES CC[N+](CC)(CC)C1=CC=CC=C1.[I-]

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 305.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Triethylphenylammonium Iodide