accessibility menu, dialog, popup

UserName

CAS RN 1010-19-1

IUPAC Name: N,N,N-triethylanilinium iodide
Synonyms: N,N,N-triethylanilinium iodide
Molecular Weight (g/mol): 305.2
Molecular Formula: C12H20IN
InChi Key: WMSWXWGJYOIACA-UHFFFAOYSA-M
SMILES: [I-].CC[N+](CC)(CC)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results